5-Methoxyanthranilsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
443340010
443340050
ACRO443340010EA
54.8
EUR
ACRO443340010
ACRO443340050
5-Methoxyanthranilsäure 97%
5-Methoxyanthranilsäure
Formel:
C₈H₉NO₃ Molecular Weight: 167.16 g/mol Schmelzpunkt: 151…151.7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016509 CAS-Nummer: 6705-03-9 |
Spezifikation Testergebnisse
Appearance | Off-white to brown to purple Powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...