1-Benzyloxy-4-brombenzol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene
442460100
442460500
ACRO442460100EA
42.3
EUR
ACRO442460100
ACRO442460500
1-Benzyloxy-4-brombenzol 97%
1-Benzyloxy-4-brombenzol
Formel:
C₁₃H₁₁BrO Molecular Weight: 263.13 Siedepunkt: 166…167 °C (4Torr) Schmelzpunkt: 62…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00028016 CAS-Nummer: 6793-92-6 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder or needles |
Infrared spectrum | Conforms |
Melting point | 59 to 64 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...