4-Amino-2-(trifluormethyl)benzonitril 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Cyano-3-trifluoromethylaniline

442470010 442470050
ACRO442470010EA 46.4 EUR
ACRO442470010 ACRO442470050
4-Amino-2-(trifluormethyl)benzonitril 97%
4-Amino-2-(trifluormethyl)benzonitril
Formel: C₈H₅F₃N₂
Molecular Weight: 186.14 g/mol
Schmelzpunkt: 144…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042155
CAS-Nummer: 654-70-6
UN: 3439
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to orange to brown Powder or crystals
Infrared spectrum Conforms
Melting point 138 to 146 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR