2-Methylisonicotinsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methylpyridine-4-carboxylic acid
443490010
443490050
ACRO443490010EA
72.4
EUR
ACRO443490010
ACRO443490050
2-Methylisonicotinsäure 97%
2-Methylisonicotinsäure
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00513435 CAS-Nummer: 4021-11-8 |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...