4-Amino-2-chlorbenzonitril 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Chlor-4-cyananilin, 4-Amino-2-chlorbenzoesäurenitril
443880100
ACRO443880100EA
75.3
EUR
ACRO443880100
4-Amino-2-chlorbenzonitril 99%
4-Amino-2-chlorbenzonitril
Formel:
C₇H₅ClN₂ Molecular Weight: 152,58 g/mol Siedepunkt: 301 °C (1013 hPa) Schmelzpunkt: 116…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035926 CAS-Nummer: 20925-27-3 UN: 3439 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to cream to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 116 to 118 °C |
Water | ≤0.5 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...