2-Amino-5-chlorphenylboronsäurepinakolester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-chlorophenyl boronic acid pinacol ester, 4-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

444470010 444470050
ACRO444470010EA 233 EUR
ACRO444470010 ACRO444470050
2-Amino-5-chlorphenylboronsäurepinakolester 97%
2-Amino-5-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,54 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06795672
CAS-Nummer: 1073371-77-3

Bestellen

Spezifikation Testergebnisse

Appearance White to pink to tan Solid
Infrared spectrum Conforms
Melting point 89 to 95 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR