4-Cyan-2-fluorphenylboronsäurepinakolester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-2-fluorobenzeneboronic acid pinacol ester

444720010 444720050
ACRO444720010EA 255 EUR
ACRO444720010 ACRO444720050
4-Cyan-2-fluorphenylboronsäurepinakolester 97%
4-Cyan-2-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247.08 g/mol
Schmelzpunkt: 83…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09998162
CAS-Nummer: 1035235-29-0

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white to yellow Solid
Infrared spectrum Conforms
Melting point 82 to 87 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR