2-Morpholino-4-pyridinboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine
445200010
445200050
ACRO445200010EA
229
EUR
ACRO445200010
ACRO445200050
2-Morpholino-4-pyridinboronsäurepinakolester 97%
2-Morpholino-4-pyridinboronsäurepinakolester
Formel:
C₁₅H₂₃BN₂O₃ Molecular Weight: 290.1700 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054039 CAS-Nummer: 888721-86-6 |
Spezifikation Testergebnisse
Appearance | White to off-white to light brown Powder or solid |
Infrared spectrum | Conforms |
Melting point | 82 to 88 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...