4-(N,N-Dimethylamino)phenylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(Dimethylamino)phenylboronic acid pinacol ester, N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
444960010
444960050
ACRO444960010EA
76.1
EUR
ACRO444960010
ACRO444960050
4-(N,N-Dimethylamino)phenylboronsäurepinakolester 97%
4-(N,N-Dimethylamino)phenylboronsäurepinakolester
Formel:
C₁₄H₂₂BNO₂ Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663854 CAS-Nummer: 171364-78-6 |
Spezifikation Testergebnisse
Appearance | Off-white to light green to tan Powder or solid |
Infrared spectrum | Conforms |
Melting point | 122 to 127 °C |
GC | ≥96 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...