Aciclovir 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, Acyclovir, 9-[(2-Hydroxyethoxy)methyl]guanine, 2-Amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6H-purin-6-one, 2-Amino-1,9-dihydro-9-[(2-hydroxyethoxy)methyl]-6H-purin-6-one, Aciclovir, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one
445240100
445240500
ACRO445240100EA
37.5
EUR
ACRO445240100
ACRO445240500
Aciclovir 98%
Acyclovir
Formel:
C₈H₁₁N₅O₃ Molecular Weight: 225,21 g/mol Schmelzpunkt: 256…260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00057880 CAS-Nummer: 59277-89-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...