4-Amino-3-nitrophenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-3-nitrophenylboronicacid

445300010 445300050
ACRO445300010EA 257 EUR
ACRO445300010 ACRO445300050
4-Amino-3-nitrophenylboronsäure 97%
4-Amino-3-nitrophenylboronsäure
Formel: C₆H₇BN₂O₄
Molecular Weight: 181,9418 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07437851
CAS-Nummer: 89466-07-9

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red Solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR