4-(Methylsulphonylamino)benzeneboronic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Methylsulphonylamino)phenylboronic acid, 4-Methylsulphonylaminophenylboronic acid, 4-(Methanesulphonamido)phenylboronic acid

445470010 445470050
ACRO445470010EA 88.7 EUR
ACRO445470010 ACRO445470050
4-(Methylsulphonylamino)benzeneboronic acid 97%
4-(Methylsulphonylamino)benzeneboronic acid
Formel: C₇H₁₀BNO₄S
Molecular Weight: 215,03 g/mol
Schmelzpunkt: 234…236 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179473
CAS-Nummer: 380430-57-9

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to cream to light brown Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR