2-(Tetrazol-5-yl)phenylboronic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(5-Tetrazolyl)phenylboronic acid, (2-(1H-Tetrazol-5-yl)phenyl)boronic acid, Boronic acid,B-[2-(2H-tetrazol-5-yl)phenyl]-, 2-(1H-Tetrazol-5-yl)phenylboronic acid
445550010
445550050
ACRO445550010EA
112
EUR
ACRO445550010
ACRO445550050
2-(Tetrazol-5-yl)phenylboronic acid 95%
2-(Tetrazol-5-yl)phenylboronic acid
Formel:
C₇H₇BN₄O₂ Molecular Weight: 189,97 g/mol Schmelzpunkt: 148…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06796325 CAS-Nummer: 155884-01-8 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Additional info | May contain varying amounts of anhydrides |
HPLC | ≥94.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...