ω-Brom-2-fluoracetophenon 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Brom-2'-fluoracetophenon, 2-Brom-1-(2-fluorphenyl)ethan-1-on, α-Brom-2-fluoracetophenon, 2-Fluorphenacylbromid, Brommethyl-o-fluorphenylketon
446670050
446670250
ACRO446670050EA
84.7
EUR
ACRO446670050
ACRO446670250
ω-Brom-2-fluoracetophenon 98%
ω-Brom-2-fluoracetophenon
Formel:
C₈H₆BrFO Molecular Weight: 217.04 g/mol Siedepunkt: 61…63 °C (.1Torr) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00278796 CAS-Nummer: 655-15-2 UN: 3265 ADR: 8 III |
Spezifikation Testergebnisse
Appearance | Yellow to green to brown Viscous liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...