2-Chloro-5-(trifluoromethyl)pyrazine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

446480010 446480050
ACRO446480010EA 129 EUR
ACRO446480010 ACRO446480050
2-Chloro-5-(trifluoromethyl)pyrazine 97%
2-Chloro-5-(trifluoromethyl)pyrazine
Formel: C₅H₂ClF₃N₂
Molecular Weight: 182.53 g/mol
Siedepunkt: 88 °C (118 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD13189478
CAS-Nummer: 799557-87-2
UN: 2922
ADR: 8 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to orange to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR