1,3,5-Triformylbenzene 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,3,5-benzenetricarbaldehyde, Trimesic formaldehyde, Benzene-1,3,5-tricarbaldehyde
446550010
446550050
ACRO446550010EA
216
EUR
ACRO446550010
ACRO446550050
1,3,5-Triformylbenzene 98%
1,3,5-Triformylbenzene
Formel:
C₉H₆O₃ Molecular Weight: 162,14 g/mol Schmelzpunkt: 156…158 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00182470 CAS-Nummer: 3163-76-6 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...