2-Methyl-2-propanyl-(3-oxocyclobutyl)carbamat 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Carbamic acid, N-(3-oxocyclobutyl)-, 1,1-dimethylethyl ester, 2-Methyl-2-propanyl (3-oxocyclobutyl)carbamate, tert-Butyl 3-oxocyclobutylcarbamate
446590010
446590050
ACRO446590010EA
202
EUR
ACRO446590010
ACRO446590050
2-Methyl-2-propanyl-(3-oxocyclobutyl)carbamat 95%
tert-Butyl-3-oxocyclobutylcarbamate
Formel:
C₉H₁₅NO₃ Molecular Weight: 185.22 g/mol Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09751872 CAS-Nummer: 154748-49-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder or solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...