3-Amino-5-ethyl-1H-pyrazol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Amino-5-ethylpyrazole

446710010 446710050
ACRO446710010EA 143 EUR
ACRO446710010 ACRO446710050
3-Amino-5-ethyl-1H-pyrazol 97%
3-Amino-5-ethyl-1H-pyrazole
Formel: C₅H₉N₃
Molecular Weight: 111.15 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08060982
CAS-Nummer: 1904-24-1

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR