2-(2-Pyrrolidinyl)pyridin 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Pyrrolidin-2-ylpyridine
446740010
446740050
ACRO446740010EA
189
EUR
ACRO446740010
ACRO446740050
2-(2-Pyrrolidinyl)pyridin 95%
2-(2-Pyrrolidinyl)pyridin
Formel:
C₉H₁₂N₂ Molecular Weight: 148,21 g/mol Siedepunkt: 82 °C (,8 mmHg) Lagertemperatur: Kühlschrank |
MDL:
MFCD01691538 CAS-Nummer: 77790-61-5 UN: 2810 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light brown Viscous liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...