3-Chlor-2-methoxy-4-pyridinboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Chloro-2-methoxypyridine-4-boronic acid, (3-Chloro-2-methoxypyridin-4-yl)boronic acid

448770010 448772500
ACRO448770010EA 167 EUR
ACRO448770010 ACRO448772500
3-Chlor-2-methoxy-4-pyridinboronsäure 95%
3-Chloro-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BClNO₃
Molecular Weight: 187.39 g/mol
Schmelzpunkt: 140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06798255
CAS-Nummer: 957060-88-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR