6-Chloro-7-iodo-7-deazapurine 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine
448850010
448850050
ACRO448850010EA
109
EUR
ACRO448850010
ACRO448850050
6-Chloro-7-iodo-7-deazapurine 96%
6-Chloro-7-iodo-7-deazapurine
Formel:
C₆H₃ClIN₃ Molecular Weight: 279.47 g/mol Schmelzpunkt: 179…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09263258 CAS-Nummer: 123148-78-7 |
Spezifikation Testergebnisse
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 173 to 180 °C |
HPLC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...