Furosemid 97+%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid

448970010 448970050
ACRO448970010EA 39.3 EUR
ACRO448970010 ACRO448970050
Furosemid 97+%
Furosemide
Formel: C₁₂H₁₁ClN₂O₅S
Molecular Weight: 330,74 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010549
CAS-Nummer: 54-31-9

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR