Cytarabin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cytosine beta-D-Arabinofuranoside, 1-β-D-Arabinofuranosylcytosine, Arabinocytidine, Ara-C, 4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Cytosine arabinoside, 4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone
449560010
449560050
449561000
ACRO449560010EA
161
EUR
ACRO449560010
ACRO449560050
ACRO449561000
Cytarabin 98%
Cytarabine
Formel:
C₉H₁₃N₃O₅ Molecular Weight: 243,22 g/mol Schmelzpunkt: 220 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00066487 CAS-Nummer: 147-94-4 |
Spezifikation Testergebnisse
Appearance | White to Beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Specific optical rotation | +155° to +162° (20°C, 589 nm) (c=1, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...