4'-Chlorobiphenyl-4-carboxaldehyde 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
[1,1'-Biphenyl]-4-carboxaldehyde, 4'-chloro-, 4'-Chlorobiphenyl-4-carbaldehyde, 4'-Chloro-4-biphenylcarbaldehyde, 4'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde
450360010
450360050
ACRO450360010EA
150
EUR
ACRO450360010
ACRO450360050
4'-Chlorobiphenyl-4-carboxaldehyde 97%
4'-Chlorobiphenyl-4-carboxaldehyde
Formel:
C₁₃H₉ClO Molecular Weight: 216,67 g/mol Schmelzpunkt: 113…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631911 CAS-Nummer: 80565-30-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 112 to 118 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...