4,5-Difluor-o-phenylendiamin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,2-Diamino-4,5-difluorbenzol, 4,5-Difluorbenzol-1,2-diamin, 4,5-Difluor-1,2-phenylendiamin

449810010 449810050 449810250
ACRO449810010EA 51.3 EUR
ACRO449810010 ACRO449810050 ACRO449810250
4,5-Difluor-o-phenylendiamin 97%
4,5-Difluor-o-phenylendiamin
Formel: C₆H₆F₂N₂
Molecular Weight: 144.12 g/mol
Schmelzpunkt: 132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061131
CAS-Nummer: 76179-40-3

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to red-brown Crystalline powder (May darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR