3-Hydroxy-4-chinolincarbonsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
449930010
449930050
ACRO449930010EA
125
EUR
ACRO449930010
ACRO449930050
3-Hydroxy-4-chinolincarbonsäure 97%
3-Hydroxy-4-chinolincarbonsäure
Formel:
C₁₀H₇NO₃ Molecular Weight: 189,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01536959 CAS-Nummer: 118-13-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with HClO4 | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...