3,5-Diiodsalicylaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Diiodo-2-hydroxybenzaldehyde

450860050 450860250
ACRO450860050EA 47.7 EUR
ACRO450860050 ACRO450860250
3,5-Diiodsalicylaldehyd 97%
3,5-Diiodsalicylaldehyd
Formel: C₇H₄I₂O₂
Molecular Weight: 373,92 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003321
CAS-Nummer: 2631-77-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Powder or solid
Infrared spectrum Conforms
Melting point 107 to 112 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR