(4-Iodphenyl)essigsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Iodophenyl)acetic acid
450900010
450900050
ACRO450900010EA
49.7
EUR
ACRO450900010
ACRO450900050
(4-Iodphenyl)essigsäure 97%
(4-Iodphenyl)essigsäure
Formel:
(C₆H₄I)CH₂COOH Molecular Weight: 262.05 g/mol Schmelzpunkt: 140…141.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082985 CAS-Nummer: 1798-06-7 |
Spezifikation Testergebnisse
Appearance | White to off-white to light brown Powder or solid |
Infrared spectrum | Conforms |
Melting point | 137 to 143 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...