2-Amino-5-methoxybenzonitril 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-methoxy-benzonitrile
451340010
451340050
ACRO451340010EA
93.3
EUR
ACRO451340010
ACRO451340050
2-Amino-5-methoxybenzonitril 95%
2-Amino-5-methoxybenzonitril
Formel:
C₈H₈N₂O Molecular Weight: 148,1600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05149280 CAS-Nummer: 23842-82-2 UN: 3439 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow to brown Powder or low melting solid |
Infrared spectrum | Conforms |
Melting point | 39 to 45 °C |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...