1-Boc-3-(bromomethyl)azetidine 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
452160010
452160050
ACRO452160010EA
203
EUR
ACRO452160010
ACRO452160050
1-Boc-3-(bromomethyl)azetidine 95%
1-Boc-3-(bromomethyl)azetidine
Formel:
C₉H₁₆BrNO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD16556174 CAS-Nummer: 253176-93-1 |
Spezifikation Testergebnisse
Appearance | White to yellow Low melting solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...