BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) 99%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS
453250010 453251000
ACRO453250010EA 332 EUR
ACRO453250010 ACRO453251000
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) 99%
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Formel: C₁₁H₁₀N₂O₆
Molecular Weight: 266,21 g/mol
Schmelzpunkt: 159…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043141
CAS-Nummer: 55750-62-4
Bestellen
Spezifikation Testergebnisse
Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥98.5 %