Cobalt(II)hydroxid 95%, rein
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cobaltdihydroxid
453892500
453890010
ACRO453892500EA
88.2
EUR
ACRO453892500
ACRO453890010
Cobalt(II)hydroxid 95%, rein
Cobalt(II)hydroxid
Formel:
H₂CoO₂ Molecular Weight: 92,95 g/mol Dichte: 3,597 g/cm³ (15 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016015 CAS-Nummer: 21041-93-0 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Pink to red Powder |
Titration excess HCl | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...