5-Thiazolcarbonsäure 98%

Lieferant: Thermo Scientific
Achtung

454540010 454540050
ACRO454540010EA 124 EUR
ACRO454540010 ACRO454540050
5-Thiazolcarbonsäure 98%
5-Thiazolecarboxylic acid
Formel: C₄H₃NO₂S
Molecular Weight: 129,1371 g/mol
Schmelzpunkt: 208…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03428539
CAS-Nummer: 14527-41-4

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR