2-Aminomethyl-1H-imidazole dihydrochloride 98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 2-Aminomethyl-1H-imidazole dihydrochloride, 2-(Aminomethyl)imidazole dihydrochloride, (1H-Imidazol-2-yl)methanamine dihydrochloride
454590010 454590050
ACRO454590010EA 66.7 EUR
ACRO454590010 ACRO454590050
2-Aminomethyl-1H-imidazole dihydrochloride 98%
2-Aminomethyl-1H-imidazole dihydrochloride
Formel: C₄H₉Cl₂N₃
Molecular Weight: 170.04 g/mol
Schmelzpunkt: 250…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738779
CAS-Nummer: 22600-77-7
Bestellen
Spezifikation Testergebnisse
Appearance White to light yellow to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 %
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure