5-Chinolincarbonsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
454760010
454760050
ACRO454760010EA
116
EUR
ACRO454760010
ACRO454760050
5-Chinolincarbonsäure 97%
5-Chinolincarbonsäure
Formel:
C₁₀H₇NO₂ Molecular Weight: 173,17 g/mol Schmelzpunkt: 337…341 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047608 CAS-Nummer: 7250-53-5 |
Spezifikation Testergebnisse
Appearance | White to off-white to light brown Solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...