2'-Desoxy-2'-fluoruridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
455000010
ACRO455000010EA
36
EUR
ACRO455000010
2'-Desoxy-2'-fluoruridin 97%
2'-Deoxy-2'-fluorouridine
Formel:
C₉H₁₁FN₂O₅ Molecular Weight: 246,19 g/mol Schmelzpunkt: 151 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01317293 CAS-Nummer: 784-71-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 147 to 153 °C |
Loss on drying | ≤1 % (1 g, 105 °C, 3 h) |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...