Cefotaxim Natriumsalz 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

454950010 454950050 454950250
ACRO454950010EA 0 EUR
ACRO454950010 ACRO454950050 ACRO454950250
Cefotaxim Natriumsalz 95%
Cefotaxime sodium salt
Formel: C₁₆H₁₇N₅O₇S₂
Molecular Weight: 477,44 g/mol
Schmelzpunkt: 201…210 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00079073
CAS-Nummer: 64485-93-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤3.0 % (105°C, 8 hrs)
HPLC ≥94 %
pH 4.5 to 6.5
Specific optical rotation +58° to +65° (20°C, 589 nm) (c=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR