Thiamphenicol 98%
Lieferant: Thermo Scientific
Synonyme:
2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulphonyl)phenyl]-2-propyl]acetamide, (Methylsulphonyl)chloramphenicol
455450010
455450050
455450250
ACRO455450010EA
20.4
EUR
ACRO455450010
ACRO455450050
ACRO455450250
Thiamphenicol 98%
Thiamphenicol
Formel:
C₁₂H₁₅Cl₂NO₅S Molecular Weight: 356,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00467983 CAS-Nummer: 15318-45-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 160 to 170 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...