Test kits, photometrisch, Nickel, LCK-Küvetten-Tests

Lieferant: Hach

LCK337 LCK537
705-3050EA 139 EUR
705-3050 705-3051
Test kits, photometrisch, Nickel, LCK-Küvetten-Tests
Umwelt-Prüfkits

Cuvette tests for water testing applications.


  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding


Haltbarkeit: 24 Monate ab Produktionsdatum


Lagerbedingungen (LCK337): 15 - 25 °C


Lagerbedingungen (LCK537): 2 - 8 °C (Kühlschranklagerung)


Standards: LCK337: DIN 38406-E11


Zubehörinformationen: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.


Bestellinformation: For total nickel digestion use crack set LCW902

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR