Transfection reagent, TransIT-TKO®
Lieferant: MIRUS
TransIT-TKO® is a broad spectrum siRNA transfection reagent that enables high efficiency siRNA delivery and knockdown of target gene expression in many cell types, including primary cells.
- Broad spectrum siRNA and plasmid DNA delivery
- Low cellular toxicity
- Reproducible results
Co-transfection of siRNA and DNA is also feasible with TransIT-TKO® and has been tested across a variety of cell types. Transfections with TransIT-TKO® do not require medium changes and can be carried out in serum-containing medium.