Transfection reagent, TransIT®-Oligo
Lieferant: MIRUS
MIR2160
MIR2164
MIR2165
MIR2166
MIRUMIR2160EA
739
EUR
MIRUMIR2160
MIRUMIR2164
MIRUMIR2165
MIRUMIR2166
Transfection reagent, TransIT®-Oligo
Transfektionsreagenzien
TransIT®-Oligo is a high efficiency, low toxicity, transfection reagent optimised for oligonucleotide delivery into a wide range of cell types.
- Unique formulation for maximising transfection performance of oligonucleotides into a wide range of cells
- Low cellular toxicity: Maintain cell density and reduce experimental bias
- Achieve high transfection efficiency in cells to ensure experimental success
Oligonucleotides tested include phosphodiester DNA, phosphothioate DNA (sDNA), phosphothioate RNA (sRNA), 2'OMe RNA, 2'OMe RNA/sDNA Chimerics, Morpholino/DNA duplexes, siRNA and miRNA. Transfections with TransIT-Oligo do not require medium changes and can be carried out in serum-containing medium.