Electroporation kits, Ingenio®
Lieferant: MIRUS
MIR50113 MIR50116 MIR50119 MIR50115 MIR50112 MIR50118
MIRUMIR50113EA 432 EUR
MIRUMIR50113 MIRUMIR50116 MIRUMIR50119 MIRUMIR50115 MIRUMIR50112 MIRUMIR50118
Electroporation kits, Ingenio®
Elektroporationssysteme Elektroporations-Kits
Ingenio® Electroporation Kits and Solution provide a universal, high efficiency, low toxicity solution for electroporation of DNA or siRNA into hard to transfect cell types.
  • Increased gene expression in several different cell types with minimal toxicity
  • Compatible with multiple conventional electroporation instruments, including Lonza Amaxa® Nucleofector®, Bio-Rad® Gene Pulser and Harvard-BTX® electroporators
  • Can be used for electroporation with both exponential decay as well as square wave forms
Ingenio® is provided a standalone solution or as part of a kit with cuvettes (0,2 and 0,4 cm) and cell droppers. The 0,2 cm cuvettes are part of the Ingenio® Electroporation Kit compatible with the Lonza-Amaxa® Nucleofector® II/2b, and the 0,4 cm cuvettes are part of the kit compatible with other electroporators, including Bio-Rad® and Harvard-BTX®.