Transfection kit, TransIT-HeLaMONSTER®
Lieferant: MIRUS
MIR2900
MIR2904
MIR2905
MIR2906
MIRUMIR2900EA
682
EUR
MIRUMIR2900
MIRUMIR2904
MIRUMIR2905
MIRUMIR2906
Transfection kit, TransIT-HeLaMONSTER®
Transfektionsreagenzien
The TransIT-HeLaMONSTER® transfection kit is specifically optimised to provide exceptional transfection efficiency of plasmid DNA in HeLa cells and cell types of associated lineage.
- High transfection efficiency (50 to 60%)
- Low cellular toxicity: Maintain cell density and reduce experimental bias
- Suitable for both transient and stable transfection
- Deliver single or multiple plasmids – ideal for specialised applications, such as gene expression and promoter analysis
Transfection with TransIT-HeLaMONSTER® kit does not require medium changes and can be carried out in serum-containing medium.
Lieferumfang: Each kit contains a base and booster reagent to maximise transfection performance in HeLa cells.