Transfection kit, TransIT®-CHO
Lieferant: MIRUS
MIR2170 MIR2174 MIR2175 MIR2176
MIRUMIR2170EA 682 EUR
MIRUMIR2170 MIRUMIR2174 MIRUMIR2175 MIRUMIR2176
Transfection kit, TransIT®-CHO
Transfektionsreagenzien
TransIT®-CHO transfection kit is specifically optimised to provide exceptional transfection efficiency of plasmid DNA in CHO cells and cell types of associated lineage.
  • High transfection efficiency (50 to 60%)
  • Low cellular toxicity: Maintain cell density and reduce experimental bias
  • Suitable for both transient and stable transfection
Transfection with TransIT-CHO kit does not require medium changes and can be carried out in serum-containing medium.
Lieferumfang: Each kit contains a base and booster reagent to maximise transfection performance in CHO cell types.