E/E 96% ≥96% (v/v)
Lieferant: KLINIPATH
|
|
Synonyme:
Ethyl alcohol
4096-9005
4096-9010
VWRK4096-9005EA
21.8
EUR
VWRK4096-9005
VWRK4096-9010
VWRK4096-9020
E/E 96% ≥96% (v/v)
Ethanol
Formel:
C₂H₅OH Molecular Weight: 46.07 g/mol Siedepunkt: 78,3 Schmelzpunkt: -117 Flammpunkt: 12 |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 UN: 1170 ADR: 3 II Merck Index: 12,03806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...