Nucleic acid labeling kits, Label IT®
Lieferant: MIRUS
MIR3100 MIR3125 MIR3200 MIR3225 MIR3300 MIR3325 MIR3400 MIR3425 MIR3600 MIR3625 MIR3725 MIR3800 MIR3825 MIR4100 MIR4125 MIR7100 MIR7125
MIRUMIR3100EA 973 EUR
MIRUMIR3100 MIRUMIR3125 MIRUMIR3200 MIRUMIR3225 MIRUMIR3300 MIRUMIR3325 MIRUMIR3400 MIRUMIR3425 MIRUMIR3600 MIRUMIR3625 MIRUMIR3725 MIRUMIR3800 MIRUMIR3825 MIRUMIR4100 MIRUMIR4125 MIRUMIR7100 MIRUMIR7125
Nucleic acid labeling kits, Label IT®
Tests Nukleinsäure-Tests
Label IT® nucleic acid labeling kits are suitable for direct non-enzymatic labeling of DNA and RNA. Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications.
  • Easily and consistently control the labeling reactions
  • Achieve high sensitivity with optimally labeled DNA and RNA
  • Do not impact hybridisation performance
All Label IT® reagent kits use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Lieferumfang: Each kit contains a Label IT® reagent, Label IT® reconstitution solution, 10X labeling buffer and solutions.