Intracellular nucleic acid localisation kits, Label IT® Tracker™
Lieferant: MIRUS
MIR7025
MIR7020
MIR7021
MIR7022
MIR7023
MIR7024
MIRUMIR7025EA
395
EUR
MIRUMIR7025
MIRUMIR7020
MIRUMIR7021
MIRUMIR7022
MIRUMIR7023
MIRUMIR7024
Intracellular nucleic acid localisation kits, Label IT® Tracker™
Tests
Nukleinsäure-Tests
Label IT® Tracker™ is designed for efficient, direct labeling of plasmid DNA for in vitro and in vivo tracking experiments.
- Efficiently label and visualise the DNA of choice
- Easily and consistently control the labeling reactions
- Optimal visualisation of DNA in cells
- Monitor both subcellular localization and reporter transgene expression following transfection of labeled (plasmid) DNA
Lieferumfang: Each kit contains Label IT® Tracker™ reagent, Label IT® reconstitution solution and 10X labeling buffer, sufficient to label 50 to 200 µg of plasmid DNA.