Ethanol 96°
Gefahr
|
Synonyme:
Ethyl alcohol
0110F5
ALCH0110F5EA
13.1
EUR
ALCH0110F5
Ethanol 96°
Ethanol
Rectified Alcohol From Molasses For Food Use
Storage temperature: min: 5 °C, max: 40 °C (recommended).
Formel:
C₂H₅OH Molecular Weight: 46,07 g/mol Dichte: >0,805 |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 UN: 1170 ADR: 3,II |
Spezifikation Testergebnisse
Alcoholic grade | > 96,0 |
Appearance | Colorless |
Odor | Neutral |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...