Endotoxin removal kit, MiraCLEAN®
Lieferant: MIRUS
MIR5910 MIR5900
MIRUMIR5910EA 306 EUR
MIRUMIR5910 MIRUMIR5900
Endotoxin removal kit, MiraCLEAN®
Reinigungsmittel Dekontaminierungsmittel
MiraCLEAN® offers a convenient and effective method for the removal of endotoxin from plasmid DNA preparations.
  • Aids in the safety and efficiency of gene delivery research
  • Removes harmful endotoxin that could cause decreased transfection efficiencies in vitro and inflammatory reactions in vivo
  • Simple phase separation protocol allows quick visualisation of recovered nucleic acid preparations