Transfection reagents, TransIT®-2020
Lieferant: MIRUS
MIR5400 MIR5405 MIR5404 MIR5406
MIRUMIR5400EA 767 EUR
MIRUMIR5400 MIRUMIR5405 MIRUMIR5404 MIRUMIR5406
Transfection reagents, TransIT®-2020
Transfektionsreagenzien
TransIT®-2020 is a high performance, animal-free, broad spectrum DNA transfection reagent.
  • Achieve high expression in many cell types, including hard to transfect cell lines and primary cells
  • Balances high efficiency nucleic acid delivery and low cellular toxicity
  • High performance with maximum compatibility